Methyl propionate
EHERC16493600
:
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
:
:88.11 g/mol
:12,06191
:79.7 °C (1013 hPa)
:–87 °C
:0.91 g/cm³ (20 °C)
:–2 °C
:Ambient
:209-060-4
:1248
:3,II
:GHS02
:GHS07
:554-12-1
:C₄H₈O₂
:MFCD00009306