Italy
Order Entry
Methyl propionate
  EHERC16493600
 :  
Methyl propionate
  EHERC16493600
 :  
83,00 €
  1 * 1 mL
  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
 :
 :88.11 g/mol
 :12,06191
 :79.7 °C (1013 hPa)
 :–87 °C
 :0.91 g/cm³ (20 °C)
 :–2 °C
 :Ambient
 :209-060-4
 :1248
 :3,II
 :GHS02
 :GHS07
 :554-12-1
 :C₄H₈O₂
 :MFCD00009306