Italy
Order Entry
Acetaldehyde analytical standard, Supelco®
  506788.
 :  Merck
 :  
undefined
Acetaldehyde analytical standard, Supelco®
  506788.
 :  Merck
 :  
64,20 €
  1 * 1 pezzi
  • Pk:
    1.000 mg
  • Seq:
    2865
  • Environmentally Preferable:
 : Ethanal
 :
 :44.05 g/mol
 :–27 °C
 :200-836-8
 :1089
 :GHS02
 :GHS08
 :GHS07
 :Refrigerator
 :75-07-0
 :CH₃CHO
 :MFCD00006991
 :20.1
 :–123 °C
 :0.78 g/cm³ (20 °C)
 :3 I
 :12,00037