(±)-2-Ethyl-1-hexanol, Supelco®
: Merck
:104-76-7
:
:CH₃(CH₂)₃CH(C₂H₅)CH₂OH
:MFCD00004746
:130.23 g/mol
:13,03842
:183 °C (1013 hPa)
:–76 °C
:0.83 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:81 °C
:Ambient
:203-234-3
:01-2119487289-20
:GHS07
- Pack typePk