Bromoform 99+% stabilised
:75-25-2
:
:Br₃CH
:MFCD00000128
:252.73 g/mol
:13,01404
:150…151 °C (760 mmHg)
:8 °C
:2.89 g/cm³
:Ambient
:200-854-6
:2515
:6.1,III
:GHS06
:GHS09
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥99.0 % (excluding stabilizer)
- Stabilizer:≤1.5 % w/w ethanol
- Pack typePk
- 220691000Glass bottle100 mL1 * 100 mL162,00 €
- 220695000Glass bottle500 mL1 * 500 mL794,00 €