Skip to Header
Skip to Main Content
Skip to Footer
/
Italy
Products
Services
Digital Solutions
Applications
Support
About
0,00 €
Order Entry
Home
Nucleic Acid Reagents
Restriction Enzymes
Restriction enzymes, NdeI, Fermentas
Restriction enzymes, NdeI, Fermentas
Restriction enzymes, NdeI, Fermentas
:
Thermo Fisher Scientific
Restriction enzymes, NdeI, Fermentas
:
Thermo Fisher Scientific
Compatible buffer:
FastDigest Green Buffer or FastDigest Buffer (colorless)
Overhang type:
5' overhang
Enzyme name:
NdeI
Concentration
Units
Pk
NdeI
NdeI
ER0581
ER0581
10 U/µl
500 units
1 KIT
1 * 1 KIT
1 * 1 KIT - 40,62 €
40,62 €
-
+
1 * 1 KIT
40,62 €
-
+
NdeI
NdeI
ER0582
ER0582
10 U/µl
2500 units
1 KIT
1 * 1 KIT
1 * 1 KIT - 176,00 €
176,00 €
-
+
1 * 1 KIT
176,00 €
-
+
NdeI
NdeI
ER0585
ER0585
10 U/µl
4000 units
1 KIT
1 * 1 KIT
1 * 1 KIT - 254,00 €
254,00 €
-
+
1 * 1 KIT
254,00 €
-
+