Stampa…
Abrasive sheet/painters sand paper, Fre-Cut™ 255P
Fornitore: 3M
Fre-Cut™
MMMA63119EA 40.9 EUR
MMMA63119
Abrasive sheet/painters sand paper, Fre-Cut™ 255P
Panni Tamponi e kit per pulizia
These abrasive sheets can be used for the intermediate and final sanding of furniture parts, varnished surfaces, kitchen counter-tops and plastic parts by hand and with portable sanding machines.
  • C-weight paper backing gives good mineral support meaning longer product life and sustained cutting action
  • Features the 3M Hookit fixing system, meaning that in conjunction with Hookit accessories, these products can be attached and reattached as many times as necessary
  • Enhanced anti-loading coating to enhance product working life
Materials: Lacquered hard & soft wood, paints, plastics, solid surface materials (e.g Corian™), metal.
Grit: 240
Mineral: Aluminium oxide
Backing: C-weight paper
Coating: Closed
Bond: Resin