Anti-Caveolin-1 Polyclonal Antibody
Catalog # BSBTPB9165
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Caveolin-1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for CAVEOLIN 1/CAV1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CAVEOLIN 1/CAV1 information: Molecular Weight: 20472 MW; Subcellular Localization: Golgi apparatus membrane; Peripheral membrane protein. Cell membrane; Peripheral membrane protein. Membrane, caveola; Peripheral membrane protein. Membrane raft. Colocalized with DPP4 in membrane rafts. Potential hairpin- like structure in the membrane. Membrane protein of caveolae; Tissue Specificity: Expressed in muscle and lung, less so in liver, brain and kidney.
Type: Primary
Antigen: Caveolin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse