Stampa…
VWR®, Sealing Mats for PCR Plates, 96 Well
Fornitore: VWR
VWR®
VWRU82006-692
VWR®, Sealing Mats for PCR Plates, 96 Well
Guarnizioni Sigilli per micropiastre
Sealing mats, film, and foil enhance full-plate sealing and analysis.
  • Pierceable
  • Create an evaporation-resistant seal to minimize sample loss
Used in medical, pharmaceutical and food industry and molecular biology.
Certificazioni: Certified free of detectable RNase, DNase, DNA, and PCR inhibitors.