Stampa…
VWR® Signature™ Genomic Filter Tips
Fornitore: Avantor
VWR® Signature™
VWRU46620-648
VWR® Signature™ Genomic Filter Tips
Puntali per pipette
These tips incorporate a robotically-inserted filter that is made from high molecular weight polyethylene.
  • For use with most popular pipettors
  • Tips are ideal for pipetting sticky proteins or rDNA
  • Large orifice to prevent the shearing of cell membranes
  • Provide complete protection from sample carry-over and help prevent pipettor contamination from aerosols or fluids
  • Samples can be completely recovered without cutting into the tip
  • Free of RNase, DNase, endotoxins, and metals
  • Autoclavable using 15 psi at 122 °C (252 °F) for 15 minutes
The filters allow the pipettor to draw by venting through a tortuous sponge-like pathway. Pores in the pathway have an average size of 10 µm, so even radionucleotides are captured before reaching the pipettor barrel. For PCR, DNA manipulation, and P32.