Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
Login
/
Register
Italy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
0
0,00 €
Order Entry
Home
Disposable Garments
Shoe covers, non woven PP
Shoe covers, non woven PP
Shoe covers, non woven PP
HOIT110177
:
Hubei Orient International
Shoe covers, non woven PP
HOIT110177
:
Hubei Orient International
165,00 €
1 * 1.000 pezzi
-
+
Shoe size:
U*
Garment application:
Laboratory
,
Food
Colour:
Blue
Pk:
1.000
About this item