- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:N-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Protein synonyms:Fatty Acid-Binding Protein 6|Intestinal Bile Acid-Binding Protein|FABP6|I-BABP|ILLBP|Gastrotropin|ILBP|I-15P|Ileal Lipid-Binding Protein|Intestinal 15 kDa Protein|GT
- Protein/peptide name:FABP6
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:16.6 kD
- Sequence:Met1-Ala128
- Formulation:Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 0.5mM DTT, 50% Glycerol, pH 8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:For Biological assays
Fatty Acid-Binding Protein 6 (FABP6) is cytoplasmic protein that binds long-chain fatty acids and other hydrophobic ligands which belongs to the calycin superfamily. FABP6 expression is restricted in the small intestine to the ileum where it is involved in the enterohepatic circulation of bile acids. FABP6 forms a beta-barrel structure that accommodates the hydrophobic ligand in its interior. Isoform 2 is expressed in colorectal adenocarcinomas and their adjacent normal mucosa (at protein level). Isoform 1 is expressed in the jejunum, ileum, cecum and ascending colon intestine. FABP6 plays a role in fatty acid uptake, transport, and metabolism. FABP6 stimulates gastric acid and pepsinogen secretion. It seems to be able to bind to bile salts and bilirubins.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag