Italy
Order Entry
Anti-PLK1 Mouse Monoclonal Antibody
  PRSI48-826
undefined
Anti-PLK1 Mouse Monoclonal Antibody
  PRSI48-826
 
694,00 €
   
  1 * 50 µG

 

  • Antigen name:
    Polo like kinase 1
  • Antigen symbol:
    PLK1
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    polo-like kinase|Serine/threonine-protein kinase 13|Polo-Like Kinase pS482/pS486/pS490,STPK13|PLK-1|polo-like kinase 1
  • Immunogen:
    Polo - Like Kinase monoclonal antibody was raised against a modified peptide of Polo - Like Kinase (Human).
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

PLK, a Polo type protein kinase, has been shown to function in mitosis. The mRNA is absent or expressed at very low levels during the G1 phase, begins to reaccumulate during the S phase, and reaches maximal levels during the G2/M phase. PLK1 may participate in targeting MPF to the nucleus during prophase and in regulating mitotic spindle function. These functions, in addition to its expression in proliferating cells, suggest that PLK1 has a role in cancer progression.

Affinity Chromatography

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Aliquot and store at 4ºC. Avoid freezing.

Type:
Antigen: PLK1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human