Stampa…

Anti-RAD51L1 Goat Polyclonal Antibody

PRSI46-262EA 429 EUR
PRSI46-262
Anti-RAD51L1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 8,000

RAD51L1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RAD51L1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RAD51L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: 0
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR