Stampa…

Anti-RAD51L3 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-261EA 435 EUR
PRSI46-261
Anti-RAD51L3 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 65 kDa band in human brain and A431 lysates at 1 μg/mL, this band was successfully blocked by incubation with the immunizing peptide.

RAD51D antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RAD51D antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RAD51L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR