Stampa…

Anti-RACGAP1 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-260EA 553 EUR
PRSI46-260
Anti-RACGAP1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in human testis lysates (calculated MW of 71.0 kDa according to NP_037409.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Toure et al., J Biol Chem 1998 Mar 13;273 (11) :6019-23; PMID: 9497316). Recommended concentration: 0.1-0.3 μg/mL. Approximately 75 kDa band observed in K562 and Jurkat lysates (calculated MW of 71 kDa according to NP_037409). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human testis, shows nuclear staining of many cells. Recommended concentration, 1 μg/mL.

RACGAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

RACGAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RACGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR