Stampa…

Anti-QKI Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-256EA 553 EUR
PRSI46-256
Anti-QKI Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 38 kDa band observed in mouse brain lysates (calculated MW of 35.8 kDa according to NP_068681.1). Recommended concentration: 0.3-1 μg/mL.

Quaking antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Quaking antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: QKI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR