Stampa…

Anti-PC Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-255EA 553 EUR
PRSI46-255
Anti-PC Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 150 kDa band observed in mouse liver lysates (calculated MW of 130 kDa according to NP_000911.2, NP_001035806.1and NP_071504.2). Recommended concentration: 0.03-0.1 μg/mL.

Pyruvate carboxylase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Pyruvate carboxylase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR