Stampa…

Anti-S100 A7 Goat Polyclonal Antibody

PRSI46-241EA 546 EUR
PRSI46-241
Anti-S100 A7 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 15 kDa band in human skin lysates after 0.3 μg/mL antibody staining.5 kDa according to NP_002954.1. The 15 kDa band was successfully blocked by incubation with the immunizing peptide.

Psoriasin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Psoriasin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: S100 A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR