Stampa…

Anti-PSD3 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-237EA 435 EUR
PRSI46-237
Anti-PSD3 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 38-40 kDa band in human brain (cerebellum and amygdala) lysates after 0.3 μg/mL antibody staining.3. The 38-40 kDa band was successfully blocked by incubation with the immunizing peptide.

PSD3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PSD3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PSD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR