Keep my session open?
Ending In 
The session is expired
La tua sessione è scaduta. Per favore rifare il login

Anti-PRPF31 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 60 kDa band observed in lysates of cell lines A431 and HeLa (calculated MW of 55.4 kDa according to NP_056444.2). Recommended concentration: 1-3 μg/mL. an additional band of unknown identity was also consistently observed at 35 kDa . This band was successfully blocked by incubation with the immunizing peptide. Also a band of 28 kDa was consistently observed, however this band was not blocked by the immunizing peptide and it is therefore a non-specific signal. We call for caution when used for other assays than Western Blot.

PRPF31 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PRPF31 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRPF31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human




 

Ordine

Espandi tutto / Chiudi tutto
Taglia Conf Codice catalogo VWR Quantità
100 µg 100 µG PRSI46-230
Codice articolo fornitore: 46-230
Codice articolo locale: PRSI46-230

Specifiche

Antigene PRPF31
Nome antigene Pre-mRNA processing Factor 31
Reattività Human
Coniugazione Unconjugated
Clonabilità Polyclonal
Host Goat
Western blot Yes
ELISA Yes
Sinomini antigene pre-mRNA processing factor 31 homolog (yeast), PRP31 pre-mRNA processing factor 31 homolog (S. cerevisiae), pre-mRNA processing factor 31 homolog, NY-BR-99, DKFZp566J153, PRP31 pre-mRNA processing factor 31 homolog (yeast), PRP31, PRPF31, RP11
Concentrazione 500 ug/mL
Immunogeno PRPF31 antibody was raised against a 15 amino acid synthetic peptide near the internal region of PRPF31.
Forma Liquid
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Seleziona tra i certificati dei lotti più recenti:

o
Confronta le alternative
Abbiamo individuato delle alternative che potrebbero farti risparmiare  per articolo.  Per confrontare gli articoli, puoi selezionare fino a 3 alternative proposte e cliccare "Confronta". Per aggiungerli al carrello inserisci una quantità e clicca "Aggiungi al carrello".

Prodotto originariamente selezionato:

Descrizione Codice catalogo Disponibilità Unità Il tuo prezzo Prezzo per Quantità