Stampa…

Anti-PENK Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-227EA 553 EUR
PRSI46-227
Anti-PENK Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 37 kDa band observed in human adrenal gland lysates (calculated MW of 30.8 kDa according to NP_006202.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Normant and Loh, Endocrinology. 1998 Apr;139 (4) :2137-45; PMID: 9529003). Recommended concentration: 1-3 μg/mL.

Proenkephalin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

ProenkepH alin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PENK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR