Stampa…

Anti-PTP4A1 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-225EA 553 EUR
PRSI46-225
Anti-PTP4A1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 21 kDa band observed in lysates of cell lines A549 and HeLa (calculated MW of 19.8 kDa according to NP_003454.1). Recommended concentration: 1-3 μg/mL.

PRL-1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PRL-1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PTP4A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR