Stampa…

Anti-PRDM11 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI46-215EA 553 EUR
PRSI46-215
Anti-PRDM11 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band observed in human lung carcinoma A549 cell line lysates (calculated MW of 58 kDa according to NP_064614.2). Recommended concentration: 2-3 μg/mL.

PRDM11 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PRDM11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PRDM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR