Stampa…

Anti-NRTN Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-962EA 553 EUR
PRSI45-962
Anti-NRTN Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 25 kDa band observed in human heart and mouse heart lysates (calculated MW of 22.4 kDa according to NP_004549.1). Recommended concentration: 0.1-0.3 μg/mL.

Neurturin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Neurturin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NRTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR