Stampa…

Anti-DPF1 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-954EA 553 EUR
PRSI45-954
Anti-DPF1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 35 kDa band observed in human cerebellum lysates (calculated MW of 37.9 according to NP_001128628.1). Recommended concentration: 1-3µg/ml

Neuro-d4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Neuro-d4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR