Stampa…

Anti-NEIL1 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-942EA 553 EUR
PRSI45-942
Anti-NEIL1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 60 kDa band in human liver and brain lysates at 1 μg/mL.7kDda according to NP_078884.1. Immunohistochemistry: In paraffin embedded human placenta shows strong staining of cytoplasm in syncytiotrophoblasts. Recommended concentration, 5-10 μg/mL.

NEIL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NEIL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NEIL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR