Stampa…

Anti-NANOG Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-932EA 553 EUR
PRSI45-932
Anti-NANOG Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:128000. Western Blot: Approximately 35 kDa band observed in nuclear HeLa lysate (calculated MW of 34.6 kDa according to NP_079141.1). Recommended concentration: 0.05-0.1 μg/mL. an additional band of approximately 37 kDa is observed and is consistent with the observations of higher molecular weight bands regularly found with antibodies from other sources. Immunohistochemistry: In paraffin embedded human testis shows nuclear staining in spermatogonia. Recommended concentration, 1-2 μg/mL.

NANOG antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NANOG antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NANOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR