Stampa…

Anti-BTBD14B Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-930EA 553 EUR
PRSI45-930
Anti-BTBD14B Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 55 kDa band observed in human bone marrow lysates (calculated MW of 57.3 kDa according to NP_443108.1). Recommended concentration: 0.5-1.5 μg/mL.

NAC1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

NAC1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BTBD14B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR