Stampa…

Anti-MYBL2 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-923EA 553 EUR
PRSI45-923
Anti-MYBL2 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 100 kDa band observed in rat stomach lysates (calculated MW of 78.8 kDa according to human NP_002457.1). Recommended concentration: 0.1-0.3 μg/mL.

MYBL2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MYBL2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MYBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR