Stampa…

Anti-MTR Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-914EA 553 EUR
PRSI45-914
Anti-MTR Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 150 kDa band seen in HeLa and Jurkat cell lysates. Recommended for use at 1-3 μg/mL.

MTR antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MTR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MTR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR