Stampa…

Anti-MORF4L1 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-901EA 553 EUR
PRSI45-901
Anti-MORF4L1 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 40 kDa band in human liver lysates and an approximately 100 kDa band in human heart lysates at 1 μg/mL. Immunohistochemistry: In paraffin embedded human uterus shows nuclear staining in the epithelial cells of the endometrial glands. Recommended concentration, 2-4 μg/mL.

MRG15 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MRG15 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MORF4L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR