Stampa…

Anti-MPP5 Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-898EA 553 EUR
PRSI45-898
Anti-MPP5 Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution >1:32,000. Western Blot: Approximately 75 kDa band observed in human brain lysate (calculated MW of 77.3 kDa according to NP_071919). Recommended concentration: 1-3 μg/mL. Immunohistochemistry: In paraffin embedded human skeletal muscle shows strong alternating staning of muscle fibres in longitudinal section. Recommended concentration, 5-10 μg/mL.

MPP5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MPP5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MPP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR