Stampa…

Anti-MGLL Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-896EA 553 EUR
PRSI45-896
Anti-MGLL Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 30 kDa band observed in human adipose and frontal cortex lysates (calculated MW of 33.3 kDa according to NP_001003794.1). In transfected HEK293 transiently expressing MGLL a band of approximately 36 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.5-1.5 μg/mL.

Monoglyceride Lipase antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Monoglyceride Lipase antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MGLL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR