Stampa…

Anti-MLPH Goat Polyclonal Antibody

Fornitore: ProSci Inc.
PRSI45-882EA 553 EUR
PRSI45-882
Anti-MLPH Goat Polyclonal Antibody
Anticorpi
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 80 kDa band observed in mouse Melanocyte extracts. Recommended for use at 0.5-2 μg/mL. Please note that this antibody may require overnight incubation of primary and high protein loads.

Melanophilin antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

MelanopH ilin antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MLPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR