Anti-E2F6 Goat Polyclonal Antibody
PRSI42-237
- Antigen name:E2F transcription Factor 6
- Antigen symbol:E2F6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MGC111545|E2F6_HUMAN.|E2F binding site modulating activity protein|Transcription factor E2F6|E2F transcription factor 6 isoform 1|EMA|E2F transcription factor 6|E2F 6
- Concentration:500 ug/mL
- Immunogen:E2F6 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of E2F6.
- Size:100 µg
- Pk:100 µG
E2F6 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
E2F6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: E2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human