Stampa…
Automated liquid handling workstation, Microlab® STAR™ Line
Fornitore: Hamilton
Microlab® STAR™
HAMISTARPLUSEA 0 EUR
HAMISTARPLUS HAMI173000 HAMISTARLET
Automated liquid handling workstation, Microlab® STAR™ Line
Automated Liquid Handling Systems
The Microlab STAR Line of workstations uses air displacement pipetting and the unique CO-RE system for forceless tip pickup and ejection. Due to the modular design, changes and upgrades to existing configurations are easy.
  • Automated intelligence with Hamilton’s air-based advanced pipetting features
  • Reduced risk of contamination for critical assays due to the combination with disposable tips
  • Less maintenance required due to no system liquid, diluters, valves or tubing
  • No dilution effect of samples
The scalability of STAR Line instruments means that the widest possible range of throughputs and budgets can be accommodated: Additional pipetting channels, a CO-RE 96, 384 Probe Head or an iSWAP plate handling arm can be fitted to existing configurations.
Deck capacity:
Microlab STAR
54 tracks (T) allow combinations of: maximum of 9 carriers (6 T) holding 5 plates or tip racks or per carrier. Multiprobe head can reach up to 7 carriers on the deck and 65mm beyond the deck (on the left side).
Microlab STARlet
30 tracks (T) allow combinations of: maximum of 30 tube carriers (1 T) holding 24 or 32 tubes per carrier maximum of 5 carriers (6 T) holding 5 tip racks or 5 plate positions per carrier
Microlab STARplus
82 tracks (T) allow combinations of: maximum of 11 carriers (6 T) holding 5 plates or tip racks per carrier plus 16 T for the waste container and on-deck components
Attenzione: * Lower weight is for configuration with 8 channels, higher weight for configuration with 96-probe head and 8 individual channels.