Stampa…
Vacuum filtration manifolds, Microsart® Combi.jet
Fornitore: Sartorius
SART16848-CJEA 2280 EUR
SART16848-CJ SART16847-CJ
Vacuum filtration manifolds, Microsart® Combi.jet
Rampe di filtrazione
The Microsart® Combi.jet is a 2-branch manifold, made of high-grade stainless steel. The manifold has been specifically designed for the use together with the new Microsart® e.jet transfer pump.
  • Create sufficient vacuum for vacuum filtration concurrently transferring the filtered liquid directly to waste
  • Easily connected and disassembled by the innovative Quick Connection technology
The stainless steel three-way valves allow independent control of vacuum and individual sterile venting of each filter holder on the system.