About this item
These VWR® pipette tips are made of clear, pre-tested PP resin and fit Rainin® LTS® pipettes.
- Fit and eject with significantly less force than most conventional pipette tips
- Ultra-fine points and super-clear resin for excellent viewing of sample
- Free of RNase, DNase, endotoxins and trace metals
- Tested for ATP
- Available in different packaging options
- Autoclavable
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Bulk: Tips are supplied in resealable PE bags (1000 tips). These packaging options are not recommended if you plan to store the tips for long periods and if warping is a concern.
Racks (96 tips per rack): These colour coded racks, with smooth rounded edges, have heavy hook hinges that are extremely durable, and an ergonomic snap latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Next Generation refills: Next Generation refill systems refill VWR and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution for waste reduction goals in the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips.
Specifications
- Catalog No:
- 613-0727
- 613-0582
- Environmentally Preferable:
- Low Manufacturing Impact, Sustainable Packaging
- Low Manufacturing Impact, Sustainable Packaging
Specifications
Related Information
Certificates
Search by Lot NumberCoC_Pipette tips for Rainin® LTS® Pipettes CoC_VWR_PIPETTE_TIPS_RACKED_STERILE CoC_VWR_PIPETTE_TIPS_REFILL_NEXT_GENERATION CoC_VWR_PIPETTE_TIPS_REFILL_NEXT_GENERATION_PP CoC_VWR_PIPETTE_TIPS_REFILL_NEXTGENERATION
VWRI613-0578_TDS_EN
Datasheet
VWR® for Liquid Handling
Product Literature
1 - 200 µl
Product Information
- Catalog #Tip styleVolumeSterileCE-IVDLengthPackedPkAvailabilityPrice
- 613-0727Rainin® LTS® style0,5 - 250 µl-+51,31 mmNext Generation refill (10 refills, 96 each)9601 * 960 pezziLocal Article Number: VWRI613-0727Specifications:
Low Manufacturing Impact Sustainable Packaging Accessories
Specifications
Environmentally PreferableLow Manufacturing Impact, Sustainable PackagingLow Manufacturing Impact Sustainable Packaging
100 - 1000 µl
Product Information
- Catalog #Tip styleVolumeSterileCE-IVDLengthPackedPkAvailabilityPrice
- 613-0582Rainin® LTS® style100 - 1000 µl-+88,90 mm6 racks, 96 each5761 * 576 pezzi51,80 €Local Article Number: VWRI613-0582Specifications:
Low Manufacturing Impact Sustainable Packaging Accessories
Specifications
Environmentally PreferableLow Manufacturing Impact, Sustainable PackagingLow Manufacturing Impact Sustainable Packaging