Potassio bromato soluzione 0.1 N, Titripur®
|
Pericolo
|
M-Clarity™ quality level = MQ100
1.60308.1000EA
42.3
EUR
1.60308.1000
Potassio bromato soluzione 0.1 N, Titripur®
Potassio bromato
Formula:
KBrO₃ Peso molecolare: 167 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00011359 CAS Number: 7758-01-2 ADR: 5.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...