Italy
Order Entry
Industrial label and sign printer, MiniMark™
 :  Brady
 
undefined
Industrial label and sign printer, MiniMark™
 :  Brady

Vinyl Tape B-7589, 100 mm

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 01
    Label printer tapes
    B-595
    MINIMARK BLACK 57MM
    Vinyl
    Permanent acrylic
    Black
       
       
    1 * 1 Roll
    1 * 1 Roll - 209,13 €
    209,13 €

     : 113215
     : BRDY113215

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 16
    Label printer tapes
    B-595
    MINIMARK BLUE 57MM
    Vinyl
    Permanent acrylic
    Blue
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113209
     : BRDY113209

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 07
    Label printer tapes
    B-595
    MINIMARK BLUE 29MM
    Vinyl
    Permanent acrylic
    Blue
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113210
     : BRDY113210

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 28
    Label printer tapes
    B-595
    MINIMARK PURPLE 29MM
    Vinyl
    Permanent acrylic
    Purple
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113216
     : BRDY113216

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 17
    Label printer tapes
    B-595
    MINIMARK PURPLE 100MM
    Vinyl
    Permanent acrylic
    Purple
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113217
     : BRDY113217

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 29
    Label printer tapes
    B-595
    MINIMARK GREY 57MM
    Vinyl
    Permanent acrylic
    Grey
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113218
     : BRDY113218

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 03
    Label printer tapes
    B-595
    MINIMARK YELLOW 29MM
    Vinyl
    Permanent acrylic
    Yellow
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113194
     : BRDY113194

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 23
    Label printer tapes
    B-595
    MINIMARK RED 100MM
    Vinyl
    Permanent acrylic
    Red
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113196
     : BRDY113196

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 13
    Label printer tapes
    B-595
    MINIMARK RED 57MM
    Vinyl
    Permanent acrylic
    Red
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113197
     : BRDY113197

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 20
    Label printer tapes
    B-595
    MINIMARK WHITE 100MM
    Vinyl
    Permanent acrylic
    White
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113182
     : BRDY113182

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 11
    Label printer tapes
    B-595
    MINIMARK WHITE 57MM
    Vinyl
    Permanent acrylic
    White
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113184
     : BRDY113184

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 02
    Label printer tapes
    B-595
    MINIMARK WHITE 29MM
    Vinyl
    Permanent acrylic
    White
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113185
     : BRDY113185

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 22
    Label printer tapes
    B-595
    MINIMARK YELLOW 100MM
    Vinyl
    Permanent acrylic
    Yellow
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113191
     : BRDY113191

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 12
    Label printer tapes
    B-595
    MINIMARK YELLOW 57MM
    Vinyl
    Permanent acrylic
    Yellow
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113193
     : BRDY113193

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 21
    Label printer tapes
    B-595
    MINIMARK CLEAR 100MM
    Vinyl
    Permanent acrylic
    Clear
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113187
     : BRDY113187

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 18
    Label printer tapes
    B-595
    MINIMARK GREY 29MM
    Vinyl
    Permanent acrylic
    Grey
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113219
     : BRDY113219

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 24
    Label printer tapes
    B-595
    MINIMARK ORANGE 100MM
    Vinyl
    Permanent acrylic
    Orange
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113200
     : BRDY113200

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 04
    Label printer tapes
    B-595
    MINIMARK RED 29MM
    Vinyl
    Permanent acrylic
    Red
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113198
     : BRDY113198

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 05
    Label printer tapes
    B-595
    MINIMARK ORANGE 29MM
    Vinyl
    Permanent acrylic
    Orange
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113202
     : BRDY113202

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 14
    Label printer tapes
    B-595
    MINIMARK ORANGE 57MM
    Vinyl
    Permanent acrylic
    Orange
       
       
    1 * 1 Roll
    1 * 1 Roll - 348,58 €
    348,58 €

     : 113201
     : BRDY113201

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 26
    Label printer tapes
    B-595
    MINIMARK BLUE 100MM
    Vinyl
    Permanent acrylic
    Blue
       
       
    1 * 1 Roll
    1 * 1 Roll - 487,98 €
    487,98 €

     : 113208
     : BRDY113208

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 06
    Label printer tapes
    B-595
    MINIMARK GREEN 29MM
    Vinyl
    Permanent acrylic
    Green
       
       
    1 * 1 Roll
    1 * 1 Roll - 278,85 €
    278,85 €

     : 113206
     : BRDY113206

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 37
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM BLACK
    Vinyl
    Permanent acrylic
    Black
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120867
     : BRDY120867

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 43
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM YELLOW
    Vinyl
    Permanent acrylic
    Yellow
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120862
     : BRDY120862

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 39
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM GREEN
    Vinyl
    Permanent acrylic
    Green
       
       
    1 * 1 Roll
    1 * 1 Roll - 361,64 €
    361,64 €

     : 120864
     : BRDY120864

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 40
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM ORANGE
    Vinyl
    Permanent acrylic
    Orange
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120866
     : BRDY120866

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 41
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM RED
    Vinyl
    Permanent acrylic
    Red
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120863
     : BRDY120863

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 38
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM BLUE
    Vinyl
    Permanent acrylic
    Blue
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120865
     : BRDY120865

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 42
    Label printer tape
    B-7569
    TAPE MINIMARK B7569 101MM WHITE
    Vinyl
    Permanent acrylic
    White
       
       
    1 * 1 Roll
    1 * 1 Roll - 415,89 €
    415,89 €

     : 120861
     : BRDY120861

Preprinted labels

 

  •  
    Seq
    Description
    Type
    Model
    Material
    Adhesive
    Printing method
    Colour
     
     
  • 25
    GHS/CLP printer labels
    B-7594
    MNK-CLP-105x155-L1-B7594-0.2
    PP
    Permanent acrylic
    White/red
       
       
    1 * 1 Roll
    1 * 1 Roll - 495,43 €
    495,43 €

     : 710575
     : BRDY710575

Thermal Transfer Ribbon

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 14
    MNK ribbon
    90 m
    Blue
    2
       
       
    1 * 2 pezzi
    1 * 2 pezzi - 205,82 €
    205,82 €

     : 710025
     : BRDY710025

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 15
    MNK ribbon
    90 m
    Green
    2
       
       
    1 * 2 pezzi
    1 * 2 pezzi - 205,82 €
    205,82 €

     : 710026
     : BRDY710026

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 05
    MNK high perfiormance ribbon
    90 m
    Black
    2 Roll
       
       
    1 * 2 Roll
    1 * 2 Roll - 225,25 €
    225,25 €

     : 739678
     : BRDY739678

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
    Price
     
     
  • 01
    MNK ribbon for B-595 vinyl
    70 m
    White
    1 Roll
    75,50
       
       
    1 * 1 Roll
    1 * 1 Roll - 136,36 €
    136,36 €

     : 710343
     : BRDY710343

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
    Price
     
     
  • 02
    MNK ribbon for B-595 vinyl
    90 m
    Blue
    1 Roll
    75,50
       
       
    1 * 1 Roll
    1 * 1 Roll - 136,36 €
    136,36 €

     : 710344
     : BRDY710344

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
    Price
     
     
  • 03
    MNK ribbon for B-595 vinyl
    90 m
    Red
    1 Roll
    75,50
       
       
    1 * 1 Roll
    1 * 1 Roll - 136,36 €
    136,36 €

     : 710345
     : BRDY710345

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 11
    MNK ribbon
    90 m
    Black
    2
       
       
    1 * 2 pezzi
    1 * 2 pezzi - 205,82 €
    205,82 €

     : 710022
     : BRDY710022

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 13
    MNK ribbon
    90 m
    Red
    2
       
       
    1 * 2 pezzi
    1 * 2 pezzi - 205,82 €
    205,82 €

     : 710024
     : BRDY710024

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
     
     
  • 12
    MNK ribbon
    90 m
    White
    2
       
       
    1 * 2 pezzi
    1 * 2 pezzi - 205,82 €
    205,82 €

     : 710023
     : BRDY710023

 

  •  
    Seq
    Description
    Length
    Colour
    Pk
    Price
     
     
  • 04
    MNK ribbon for B-595 vinyl
    90 m
    Green
    1 Roll
    75,50
       
       
    1 * 1 Roll
    1 * 1 Roll - 136,36 €
    136,36 €

     : 710346
     : BRDY710346