Rubber cement, fixogum
Supplier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Fixogum
ICNA11FIXO0125EA 76.7 GBP
ICNA11FIXO0125
Rubber cement, fixogum
Adhesives
Fixogum Rubber Cement is made for use in the formation of an air tight seal around the perimeter of the glass coverslip during probe hybridisation for FISH.
Order Now
Learn more