These incubating orbital shakers are available with and without refrigeration, R denotes refrigeration. Designed to maximise valuable laboratory space the units can be stacked up to three units high. The door glides up and out of the way saving space, the slide out platform gives easy access to all samples. A pull-out service module enables convenient access to all electronics and heating/cooling components without having to disturb the stacked units. Intuitive programming allows multiple parameter changes to speed, optional photosynthetic lights and optional UV light decontamination on a timed basis (R models only). There is an interior light to assist viewing and the double pane thermal glass gives visibility and insulation. These high capacity shakers can accommodate flasks up to five litres, an RS232 serial port provides two-way communication for data logging and unit control.
- Large easy to read displays shows set point and actual values
- Integral timer, 0,1 to 99,9 h, shows elapsed time or counts down, automatic switch-off and audible alarm when program is complete
- Microprocessor controller maintains precise temperature, stores programs for repeat use and recalls last set points, even when unit has been shut off. Will also restart if power is interrupted
- Triple eccentric counterbalanced drive with brushless motor ensures uniform motion regardless of where the sample is on the platform even when fully loaded and operating at top speed
- Air circulation system ensures rapid temperature equilibration and uniformity, multi-function reservoir humidifies the chamber to minimise sample evaporation
Informazioni per l' accessorio: Innova 44 and 44R shakers accommodate a maximum of 91×50, 39×125, 30×250, 24×500, 14×1000, 8×2000, 6×2800, 6×4000 or 6×5000 ml flasks using the universal platform, and 60×125, 40×250, 24×500, 15×1000, 12×2000, 6×2800 or 6×4000 ml flasks using the dedicated platforms.
Informazioni per l'ordine: Supplied with or without platforms, required accessories must be ordered separately. Factory fitted options such as remote alarm output, 12-port gassing manifold and ultraviolet germicidal light or programmable photosynthetic light bank for the 44R are available. Please contact your local VWR office before placing your order as options can not be retrofitted.