Anti-GAD1/GAD2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-13263R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:Glutamate decarboxylase 1 (brain 67kDa) And glutamate decarboxylase 2
- Antigen symbol:GAD1/GAD2
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:2572,2571
- Antigen synonyms:GAD 65|GAD 1|Glutamate decarboxylase 2|Glutamate decarboxylase 65 kDa isoform|GAD-65|GAD|GAD1|GAD 2|GAD-2|GAD-1|GAD2|Glutamate decarboxylase 1|65 kDa glutamic acid decarboxylase|GAD67|Glutamate decarboxylase 67 kDa isoform|67 kDa glutamic acid decarboxylase|GAD 67
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:370-420/585
- Purification:Purified by Protein A
- Pk:100 µl
This gene encodes one of several forms of glutamic acid decarboxylase, identified as a major autoantigen in insulin-dependent diabetes. The enzyme encoded is responsible for catalyzing the production of gamma aminobutyric acid from L glutamic acid. A pathogenic role for this enzyme has been identified in the human pancreas since it has been identified as an autoantibody and an autoreactive T cell target in insulin dependent diabetes. This gene may also play a role in the stiff man syndrome.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GAD65 + GAD67
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat