Stampa…

Anti-OR5T1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fornitore: Bioss
BOSSBS-11626R-A350EA 858 EUR
BOSSBS-11626R-A350
Anti-OR5T1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
OR5T1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OR5T1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR