Stampa…

Anti-GNAT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-11560R-A647EA 858 EUR
BOSSBS-11560R-A647
Anti-GNAT2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
Transducin is a 3-subunit guanine nucleotide-binding protein (G protein) which stimulates the coupling of rhodopsin and cGMP-phoshodiesterase during visual impulses. The transducin alpha subunits in rods and cones are encoded by separate genes. This gene encodes the alpha subunit in cones.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNAT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR