Stampa…

Anti-PTPRN Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-11532R-CY3EA 858 EUR
BOSSBS-11532R-CY3
Anti-PTPRN Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
Implicated in neuroendocrine secretory processes. May be involved in processes specific for neurosecretory granules, such as their biogenesis, trafficking or regulated exocytosis or may have a general role in neuroendocrine functions. Seems to lack intrinsic enzyme activity. May play a role in the regulation of secretory granules via its interaction with SNTB2.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TPIA2/PTPRN
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR