Stampa…

Anti-SORCS3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-11503R-A647EA 858 EUR
BOSSBS-11503R-A647
Anti-SORCS3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
This gene encodes one family member of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins. The VPS10 domain name comes from the yeast carboxypeptidase Y sorting receptor Vps10 protein. Members of this gene family are large with many exons but the CDS lengths are usually less than 3700 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. These genes are strongly expressed in the central nervous system. Two of the five family members (sortilin and sortilin-related receptor) are synthesized as preproproteins; it is not yet known if this encoded protein is also a preproprotein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SORCS3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR