Stampa…

Anti-GBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BOSSBS-11289R-A488EA 858 EUR
BOSSBS-11289R-A488
Anti-GBP2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
Hydrolyzes GTP to GMP in two consecutive cleavage reactions. Exhibits antiviral activity against influenza virus. Promote oxidative killing and deliver antimicrobial peptides to autophagolysosomes, providing broad host protection against different pathogen classes (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GBP2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR