Stampa…

Anti-CADPS2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Bioss
BOSSBS-11500R-HRPEA 858 EUR
BOSSBS-11500R-HRP
Anti-CADPS2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Calcium-dependent secretion activators (CAPS-1 and CAPS-2) are calcium-binding proteins that direct neurotransmitter and neuropeptide-filled vesicles to the cell membrane for secretory granule exocytosis. Both CAPS-1 and CAPS-2 are expressed primarily in the brain where they regulate the secretion of various substances. The CAPS proteins contain a PH domain that is essential for regulation of exocytosis, as well as regulation of phospholipid binding. Through their regulation of neurotrophin release from granule cells, CAPS proteins help to regulate cell fate during neuronal development. CAPS-1 is thought to regulate catecholamine release from neuronal cells, while CAPS-2 is thought to regulate release of both brain-derived neurotrophic factor and neurotrophin-3 from granule cells. Defects in the genes encoding CAPS-1 and CAPS-2 are implicated in impaired cerebral development and autism.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CADPS2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR