Stampa…

Anti-EDA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-1149R-A555EA 866 EUR
BOSSBS-1149R-A555
Anti-EDA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
Seems to be involved in epithelial-mesenchymal signaling during morphogenesis of ectodermal organs. Isoform 1 binds only to the receptor EDAR, while isoform 3 binds exclusively to the receptor XEDAR.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR